Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Hydroxyoctanophenone, 99%

$155.20 - $155.20

Chemical Identifiers

Chemical Name or Material 4'-Hydroxyoctanophenone
CAS 2589-73-3
Molecular Formula C14H20O2
Formula Weight 220.31
MDL Number MFCD00082694
Beilstein 1954746
Synonym Heptyl 4-hydroxyphenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1166206

  SDS
alfa aesar™
L1166206
5g Each for $155.20
Description & Specifications

Specifications

Formula Weight 220.31
CAS 2589-73-3
Chemical Name or Material 4'-Hydroxyoctanophenone
Assay 99%
Molecular Formula C14H20O2
Beilstein 1954746
Quantity 5g
MDL Number MFCD00082694
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Heptyl 4-hydroxyphenyl ketone
Melting Point 62° to 65°C
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.