missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Hydroxyoctanophenone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1166206

 View more versions of this product

Catalog No. AAL1166206



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Hydroxyoctanophenone
62°C to 65°C
MFCD00082694
1954746
GPDYSJOGSNWMDZ-UHFFFAOYSA-N
1-(4-hydroxyphenyl)octan-1-one
75767
99%
2589-73-3
C14H20O2
5 g
4-octanoylphenol, 1-4-hydroxyphenyl octan-1-one, 4'-hydroxyoctanophenone, 1-octanone, 1-4-hydroxyphenyl, octanophenone, 4'-hydroxy, unii-e2e77z6duk, e2e77z6duk, 1-4-hydroxyphenyl-1-octanone
CCCCCCCC(=O)C1=CC=C(O)C=C1
220.31
220.31

Chemical Identifiers

2589-73-3
220.31
GPDYSJOGSNWMDZ-UHFFFAOYSA-N
75767
CCCCCCCC(=O)C1=CC=C(O)C=C1
C14H20O2
MFCD00082694
4-octanoylphenol, 1-4-hydroxyphenyl octan-1-one, 4'-hydroxyoctanophenone, 1-octanone, 1-4-hydroxyphenyl, octanophenone, 4'-hydroxy, unii-e2e77z6duk, e2e77z6duk, 1-4-hydroxyphenyl-1-octanone
1-(4-hydroxyphenyl)octan-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-979-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only