Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Hydroxynonanophenone, 96%

$93.27 - $320.95

Chemical Identifiers

Chemical Name or Material 4'-Hydroxynonanophenone
CAS 14392-69-9
Molecular Formula C15H22O2
Formula Weight 234.34
MDL Number MFCD00020174
Beilstein 2103401
Synonym 4-n-Nonanoylphenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0229306

  SDS
alfa aesar™
L0229306
5g Each for $93.27

AAL0229314

  SDS
alfa aesar™
L0229314
25g Each for $320.95
Description & Specifications

Specifications

Formula Weight 234.34
Chemical Name or Material 4'-Hydroxynonanophenone
MDL Number MFCD00020174
Assay 96%
Health Hazard 1 Warning
Molecular Formula C15H22O2
Synonym 4-n-Nonanoylphenol
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 2103401
Quantity 5g
Melting Point 53° to 57°C
CAS 14392-69-9
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.