missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Hydroxymethyl)pyridine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 166111000

 View more versions of this product

Catalog No. AC166111000



Description

Description

Specifications

Specifications

4-(Hydroxymethyl)pyridine
Authentic
107°C to 110°C (1.0mmHg)
52°C to 53°C
99%
98.5
98.5% min. (GC)
(C5H4N)CH2OH
4-hydroxymethyl pyridine, 4-hydroxymethylpyridine, 4-picolyl alcohol, 4-pyridinecarbinol, 4-pyridinemethanol, 4-pyridinylmethanol, 4-pyridylcarbinol, 4-pyridylmethanol, pyridin-4-yl methanol, pyridine-4-methanol
PTMBWNZJOQBTBK-UHFFFAOYSA-N
pyridin-4-ylmethanol
11472
Crystals, Crystalline Powder or Crystalline Mass
148°C
Glass bottle
White to Yellow
100g
586-95-8
100.0
C6H7NO
MFCD00006442
Solubility in water: soluble
C1=CN=CC=C1CO
109.13
109.13
99%

Chemical Identifiers

586-95-8
109.13
PTMBWNZJOQBTBK-UHFFFAOYSA-N
11472
C1=CN=CC=C1CO
C6H7NO
MFCD00006442
4-hydroxymethyl pyridine, 4-hydroxymethylpyridine, 4-picolyl alcohol, 4-pyridinecarbinol, 4-pyridinemethanol, 4-pyridinylmethanol, 4-pyridylcarbinol, 4-pyridylmethanol, pyridin-4-yl methanol, pyridine-4-methanol
pyridin-4-ylmethanol
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-590-6

RTECSNumber : UT4691000

TSCA : TSCA

SDS Documents