Please login to your online account to display your discounted pricing

4-(Hydroxymethyl)pyridine, 99%, ACROS Organics™

$195.75 - $195.75

Chemical Identifiers

Chemical Name or Material 4-(Hydroxymethyl)pyridine
Name Note 99%
CAS 586-95-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H7NO
Linear Formula (C5H4N)CH2OH
Formula Weight 109.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166110250

  SDS
fisher scientific acros
166110250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC166111000

  SDS
thermo fisher scientific
166111000
Glass bottle 100g Each for $195.75
Description & Specifications

Specifications

Physical Form Crystals, Crystalline Powder or Crystalline Mass
Assay ≥98.5%
Formula Weight 109.13
Quantity 25g
Color White to yellow
EINECS Number 209-590-6
TSCA TSCA
RTECS Number UT4691000
Melting Point 53° to 58°C
CAS Max % 100.0
Packaging Glass bottle
Synonym 4-(Hydroxymethyl)pyridine
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H7NO
Linear Formula (C5H4N)CH2OH
MDL Number MFCD00006442
CAS 586-95-8
Health Hazard 1 HYGROSCOPIC
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-(Hydroxymethyl)pyridine
Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.