Please login to your online account to display your discounted pricing

4-(Hydroxymethyl)phenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Hydroxymethyl)phenylboronic acid
CAS 59016-93-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H9BO3
Formula Weight 151.96
MDL Number MFCD00792672
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC369470010

  SDS
thermo fisher scientific
AC369470010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 151.96
Quantity 1g
Color White to slightly yellow
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-(Hydroxymethyl)phenylboronic acid
Assay Percent Range 97%
Molecular Formula C7H9BO3
MDL Number MFCD00792672
CAS 59016-93-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C
Infrared Spectrum Authentic