Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Hydroxymethyl)phenoxyacetic acid, 98+%

$53.04 - $120.39

Chemical Identifiers

Chemical Name or Material 4-(Hydroxymethyl)phenoxyacetic acid
CAS 68858-21-9
Molecular Formula C9H10O4
Formula Weight 182.18
MDL Number MFCD00057827
Beilstein 5260336
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1572103

  SDS
alfa aesar™
L1572103
1g Each for $120.39

AAL15721MD

  SDS
alfa aesar™
L15721MD
250mg Each for $53.04
Description & Specifications

Specifications

Formula Weight 182.18
Chemical Name or Material 4-(Hydroxymethyl)phenoxyacetic acid
Assay ≥98%
MDL Number MFCD00057827
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 5260336
Health Hazard 1 Warning
Molecular Formula C9H10O4
Melting Point 110° to 113°C
Quantity 1g
Recommended Storage 2° to 8°C
CAS 68858-21-9
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C