missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Hydroxymethyl)benzoic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2218314

 View more versions of this product

Catalog No. AAB2218314


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Hydroxymethyl)benzoic acid
179°C to 183°C
C8H8O3
25 g
4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd
OCC1=CC=C(C=C1)C(O)=O
152.15
152.15
3006-96-0
Odorless
MFCD00017598
2690023
WWYFPDXEIFBNKE-UHFFFAOYSA-N
4-(hydroxymethyl)benzoic acid
76360
≥98%

Chemical Identifiers

3006-96-0
152.15
WWYFPDXEIFBNKE-UHFFFAOYSA-N
76360
OCC1=CC=C(C=C1)C(O)=O
C8H8O3
MFCD00017598
4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd
4-(hydroxymethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only