missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxymethyl-5-methylimidazole, TCI America™

$137.50 - $348.34

Chemical Identifiers

CAS 29636-87-1
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD00792476
InChI Key AXJZCJSXNZZMDU-UHFFFAOYSA-N
PubChem CID 122433
IUPAC Name (5-methyl-1H-imidazol-4-yl)methanol
SMILES CC1=C(N=CN1)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09765G View Documents TCI America
H09765G
5g Each for $137.50
 
H097625G View Documents TCI America
H097625G
25g Each for $348.34
 
Specifications

Chemical Identifiers

29636-87-1
112.132
AXJZCJSXNZZMDU-UHFFFAOYSA-N
(5-methyl-1H-imidazol-4-yl)methanol
C5H8N2O
MFCD00792476
122433
CC1=C(N=CN1)CO

Specifications

White-Yellow
5g
C5H8N2O
AXJZCJSXNZZMDU-UHFFFAOYSA-N
(5-methyl-1H-imidazol-4-yl)methanol
122433
Crystal-Powder at 20°C
4-Hydroxymethyl-5-methylimidazole
136°C
29636-87-1
MFCD00792476
CC1=C(N=CN1)CO
112.132
112.13
≥96.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : Yes