missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxymethyl-4-methyl-1-phenyl-3-pyrazolidone, 95%

$48.76 - $139.73

Chemical Identifiers

CAS 13047-13-7
Molecular Formula C11H14N2O2
Formula Weight 206.25
MDL Number MFCD00055653
Chemical Name or Material 4-Hydroxymethyl-4-methyl-1-phenyl-3-pyrazolidone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1935014 SDS Alfa Aesar™
A1935014
25g Each for $48.76
AAA1935022 SDS Alfa Aesar™
A1935022
100g Each for $139.73
Specifications Description & Specifications

Chemical Identifiers

13047-13-7
206.25
4-Hydroxymethyl-4-methyl-1-phenyl-3-pyrazolidone
C11H14N2O2
MFCD00055653

Specifications

>110°C (230°F)
25g
C11H14N2O2
MFCD00055653
95%
124° to 126°C
13047-13-7
206.25
4-Hydroxymethyl-4-methyl-1-phenyl-3-pyrazolidone

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA