missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxyindole, TCI America™

$73.33 - $261.25

Chemical Identifiers

CAS 2380-94-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005667
InChI Key NLMQHXUGJIAKTH-UHFFFAOYSA-N
Synonym 4-hydroxyindole, indol-4-ol, 4-indolol, hydroxyindole, 4-hydroxy-1h-indole, 1h-indole-4-ol, 4-hydroxy indole, unii-1w4vd9085v, hydroxyindoles, 4-hydroxy-indole
PubChem CID 75421
ChEBI CHEBI:24702
IUPAC Name 1H-indol-4-ol
SMILES C1=CC2=C(C=CN2)C(=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H058225G View Documents TCI America
H058225G
25g Each for $261.25
 
H05825G View Documents TCI America
H05825G
5g Each for $73.33
 
Specifications

Chemical Identifiers

2380-94-1
133.15
NLMQHXUGJIAKTH-UHFFFAOYSA-N
75421
1H-indol-4-ol
C8H7NO
MFCD00005667
4-hydroxyindole, indol-4-ol, 4-indolol, hydroxyindole, 4-hydroxy-1h-indole, 1h-indole-4-ol, 4-hydroxy indole, unii-1w4vd9085v, hydroxyindoles, 4-hydroxy-indole
CHEBI:24702
C1=CC2=C(C=CN2)C(=C1)O

Specifications

Undesignated
25g
C8H7NO
4-hydroxyindole, indol-4-ol, 4-indolol, hydroxyindole, 4-hydroxy-1h-indole, 1h-indole-4-ol, 4-hydroxy indole, unii-1w4vd9085v, hydroxyindoles, 4-hydroxy-indole
C1=CC2=C(C=CN2)C(=C1)O
133.15
CHEBI:24702
Crystal-Powder at 20°C
4-Hydroxyindole
99°C
2380-94-1
MFCD00005667
NLMQHXUGJIAKTH-UHFFFAOYSA-N
1H-indol-4-ol
75421
133.15
≥99.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No