Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxyindole-6-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 4-Hydroxyindole-6-carboxylic acid
CAS 40990-52-1
Formula Weight 177.16
MDL Number MFCD04973988
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6323603

 
alfa aesar™
H6323603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63236MD

 
alfa aesar™
H63236MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.16
Chemical Name or Material 4-Hydroxyindole-6-carboxylic acid
Physical Form Solid
Assay 97%
MDL Number MFCD04973988
CAS 40990-52-1
Storage Note 1 2° to 8°C
Color Pale brown to brown
Quantity 1g
Safety and Handling

2° to 8°C