missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Hydroxyhexanophenone, TCI America™

$149.72 - $360.56

Chemical Identifiers

CAS 2589-72-2
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00671540
InChI Key YDGNMJZDWLQUTE-UHFFFAOYSA-N
Synonym Amyl 4-Hydroxyphenyl Ketone, 4′-Hydroxycaprophenone
PubChem CID 345110
IUPAC Name 1-(4-hydroxyphenyl)hexan-1-one
SMILES CCCCCC(=O)C1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08845G View Documents TCI America
H08845G
5g Each for $149.72
 
H088425G View Documents TCI America
H088425G
25g Each for $360.56
 
Specifications

Chemical Identifiers

2589-72-2
192.258
YDGNMJZDWLQUTE-UHFFFAOYSA-N
345110
CCCCCC(=O)C1=CC=C(C=C1)O
C12H16O2
MFCD00671540
Amyl 4-Hydroxyphenyl Ketone, 4′-Hydroxycaprophenone
1-(4-hydroxyphenyl)hexan-1-one

Specifications

White-Yellow
5g
C12H16O2
Amyl 4-Hydroxyphenyl Ketone, 4′C-Hydroxycaprophenone
CCCCCC(=O)C1=CC=C(C=C1)O
192.258
192.26
≥99.0% (GC)
63°C
2589-72-2
MFCD00671540
YDGNMJZDWLQUTE-UHFFFAOYSA-N
1-(4-hydroxyphenyl)hexan-1-one
345110
Crystal-Powder at 20°C
4′-Hydroxyhexanophenone
Safety and Handling

Safety and Handling

TSCA : No