Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Hydroxyflavanone, 98%

$91.73 - $320.18

Chemical Identifiers

Chemical Name or Material 4'-Hydroxyflavanone
CAS 6515-37-3
Molecular Formula C15H12O3
Formula Weight 240.25
MDL Number MFCD00017705
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2482903

 
alfa aesar™
B2482903
1g Each for $91.73

AAB2482906

 
alfa aesar™
B2482906
5g Each for $320.18
Description & Specifications

Specifications

Formula Weight 240.25
Chemical Name or Material 4'-Hydroxyflavanone
Assay 98%
Molecular Formula C15H12O3
MDL Number MFCD00017705
CAS 6515-37-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈182°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.