Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Hydroxydiclofenac

A Cytochrome P450 2C9 (CYP2C9) metabolite of diclofenac

Chemical Identifiers

Chemical Name or Material 4'-Hydroxydiclofenac
CAS 64118-84-9
Formula Weight 312.15
MDL Number MFCD01671980
UN Number UN2811
Beilstein 4198042
Synonym [2-(2,6-Dichloro-4-hydroxy-phenylamino)phenyl]acetic acid; 4'-OHD
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64095LB0

  SDS
alfa aesar™
J64095LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 312.15
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4'-Hydroxydiclofenac
Synonym [2-(2,6-Dichloro-4-hydroxy-phenylamino)phenyl]acetic acid; 4'-OHD
MDL Number MFCD01671980
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Beilstein 4198042
Solubility Information Soluble in DMSO, ethanol or methanol
RTECS Number AG6542800
Physical Form Powder
Color Pale yellow
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Storage Note 1 -30° to -10°C
Quantity 5mg
CAS 64118-84-9
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

-30° to -10°C