Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxycyclohexanone ethylene acetal, 90+%

$111.95 - $111.95

Chemical Identifiers

Chemical Name or Material 4-Hydroxycyclohexanone ethylene acetal
CAS 22428-87-1
Molecular Formula C8H14O3
Formula Weight 158.19
MDL Number MFCD00067003
Synonym 1,4-Dioxaspiro[4,5]decan-8-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2603703

  SDS
alfa aesar™
H2603703
1g Each for $111.95
Description & Specifications

Specifications

Formula Weight 158.19
Synonym 1,4-Dioxaspiro[4,5]decan-8-ol
Quantity 1g
Chemical Name or Material 4-Hydroxycyclohexanone ethylene acetal
Assay ≥90%
Molecular Formula C8H14O3
MDL Number MFCD00067003
CAS 22428-87-1