missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxycyclohexanecarboxylic Acid (cis- and trans- mixture), TCI America™

$265.84 - $1,714.20

Chemical Identifiers

CAS 17419-81-7
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00077974
InChI Key HCFRWBBJISAZNK-UHFFFAOYSA-N
PubChem CID 151138
IUPAC Name 4-hydroxycyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H096025G View Documents TCI America
H096025G
25g Each for $265.84
 
H0960250G View Documents TCI America
H0960250G
250g Each for $1,714.20
 
Specifications

Chemical Identifiers

17419-81-7
144.17
HCFRWBBJISAZNK-UHFFFAOYSA-N
4-hydroxycyclohexane-1-carboxylic acid
C7H12O3
MFCD00077974
151138
C1CC(CCC1C(=O)O)O

Specifications

White
17419-81-7
MFCD00077974
C1CC(CCC1C(=O)O)O
144.17
144.17
≥98.0% (GC,T)
25g
C7H12O3
HCFRWBBJISAZNK-UHFFFAOYSA-N
4-hydroxycyclohexane-1-carboxylic acid
151138
Crystal-Powder at 20°C
4-Hydroxycyclohexanecarboxylic Acid (cis- and trans- mixture)
Safety and Handling

Safety and Handling

TSCA : No