missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxycoumarin, 98%, ACROS Organics™

$22.09 - $269.30

Chemical Identifiers

CAS 1076-38-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006856
InChI Key OWBBAPRUYLEWRR-UHFFFAOYSA-N
Synonym 2h-1-benzopyran-2-one, 4-hydroxy, 4-coumarinol, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-2h-chromen-2-one, 4-hydroxy-chromen-2-one, 4-hydroxycoumarin, benzotetronic acid, coumarin, 4-hydroxy, unii-x954zll2rd
PubChem CID 54682930
ChEBI CHEBI:40070
IUPAC Name 4-hydroxychromen-2-one
SMILES OC1=CC(=O)C2=CC=CC=C2O1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC121100050
SDS View Documents
Fisher Scientific Acros
121100050
5g Glass bottle
Each for $22.09
 
AC121101000
SDS View Documents
Thermo Fisher Scientific
121101000
100g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC121105000
SDS View Documents
Thermo Fisher Scientific
121105000
500g Plastic bottle
Each for $269.30
 
Description

Description

Specifications

Chemical Identifiers

1076-38-6
162.14
OWBBAPRUYLEWRR-UHFFFAOYSA-N
54682930
4-hydroxychromen-2-one
C9H6O3
MFCD00006856
2h-1-benzopyran-2-one, 4-hydroxy, 4-coumarinol, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-2h-chromen-2-one, 4-hydroxy-chromen-2-one, 4-hydroxycoumarin, benzotetronic acid, coumarin, 4-hydroxy, unii-x954zll2rd
CHEBI:40070
OC1=CC(=O)C2=CC=CC=C2O1

Specifications

Authentic
Glass bottle
Colorless to Yellow
5g
97.5
98%
MFCD00006856
2h-1-benzopyran-2-one, 4-hydroxy, 4-coumarinol, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-2h-chromen-2-one, 4-hydroxy-chromen-2-one, 4-hydroxycoumarin, benzotetronic acid, coumarin, 4-hydroxy, unii-x954zll2rd
OWBBAPRUYLEWRR-UHFFFAOYSA-N
4-hydroxychromen-2-one
54682930
162.14
98%
0.5% max.
0.25% max.
210°C to 215°C
1076-38-6
100.0
C9H6O3
17, 488
Solubility in water: practically insoluble in water. Other solubilities: soluble in hot water,soluble in alcohol,acetone and ether
OC1=CC(=O)C2=CC=CC=C2O1
162.14
CHEBI:40070
Liquid
4-Hydroxycoumarin, 98%
Safety and Handling

Safety and Handling

EINECSNumber : 214-060-2

RTECSNumber : DJ3100000

TSCA : TSCA

SDS
Product Certifications