missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxycoumarin, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1439436

 View more versions of this product

Catalog No. AAA1439436


Description

Description

4-Hydroxycoumarin is involved in annulation reactions, due to the relatively high acidity of the C-H bond at the 3-position: a three-component reaction with isocyanides and dialkyl acetylene dicarboxylates affords annulated 4H-pyrans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxycoumarin is involved in annulation reactions, due to the relatively high acidity of the C-H bond at the 3-position: a three-component reaction with isocyanides and dialkyl acetylene dicarboxylates affords annulated 4H-pyrans.

Solubility
Soluble in ethanol and dimethyl formamide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

4-Hydroxycoumarin
∼213°C (decomposition)
C9H6O3
500 g
4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
OWBBAPRUYLEWRR-UHFFFAOYSA-N
4-hydroxychromen-2-one
54682930
162.14
1076-38-6
Odorless
MFCD00006856
129768
Soluble in ethanol and dimethyl formamide. Insoluble in water.
OC1=CC(=O)C2=CC=CC=C2O1
162.14
CHEBI:40070
≥98%

Chemical Identifiers

1076-38-6
162.14
OWBBAPRUYLEWRR-UHFFFAOYSA-N
54682930
4-hydroxychromen-2-one
C9H6O3
MFCD00006856
4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
CHEBI:40070
OC1=CC(=O)C2=CC=CC=C2O1
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 214-060-2

RTECSNumber : DJ3100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only