missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxychalcone, 97%

Manufacturer:  Alfa Aesar™ A1530722

 View more versions of this product

Catalog No. AAA1530722



Description

Description

Applications
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.
Specifications

Specifications

4-Hydroxychalcone
100g
C15H12O2
2049155
PWWCDTYUYPOAIU-DHZHZOJOSA-N
(E)-3-(4-hydroxyphenyl)-1-phenylprop-2-en-1-one
5282361
224.26
183°C to 185°C
20426-12-4
MFCD00016488
4-hydroxychalcone, 4-hydroxy chalcone, 2e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, unii-jo97q47vbu, jo97q47vbu, 3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, e-3-4-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 3-4-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, ccris 2228
C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)O
224.259
CHEBI:34423
97%

Chemical Identifiers

20426-12-4
224.259
PWWCDTYUYPOAIU-DHZHZOJOSA-N
5282361
(E)-3-(4-hydroxyphenyl)-1-phenylprop-2-en-1-one
C15H12O2
MFCD00016488
4-hydroxychalcone, 4-hydroxy chalcone, 2e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, unii-jo97q47vbu, jo97q47vbu, 3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, e-3-4-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 3-4-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, ccris 2228
CHEBI:34423
C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UD1675000

Documents