Please login to your online account to display your discounted pricing

4-Hydroxycarbazole, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC366960050

 View more versions of this product

Catalog No. AC366960050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Color Tan to cacao
Formula Weight 183.21
Physical Form Powder
Quantity 5g
Melting Point 163° to 168°C
CAS Max % 100.0
CAS Min % 98.0
Assay Percent Range 98+%
Molecular Formula C12H9NO
MDL Number MFCD02178385
Chemical Name or Material 4-Hydroxycarbazole
CAS 52602-39-8
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Hydroxycarbazole
CAS 52602-39-8
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C12H9NO
Formula Weight 183.21
MDL Number MFCD02178385
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents