missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Hydroxybenzyl)phosphonic Acid, TCI America™

$87.08 - $262.78

Chemical Identifiers

CAS 90001-07-3
Molecular Formula C7H9O4P
Molecular Weight (g/mol) 188.119
MDL Number MFCD16877729
InChI Key ZHBRSHSRMYZHLS-UHFFFAOYSA-N
PubChem CID 12029255
IUPAC Name (4-hydroxyphenyl)methylphosphonic acid
SMILES C1=CC(=CC=C1CP(=O)(O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H1558200MG View Documents TCI America
H1558200MG
200mg Each for $87.08
 
H15581G View Documents TCI America
H15581G
1g Each for $262.78
 
Specifications

Chemical Identifiers

90001-07-3
188.119
ZHBRSHSRMYZHLS-UHFFFAOYSA-N
(4-hydroxyphenyl)methylphosphonic acid
C7H9O4P
MFCD16877729
12029255
C1=CC(=CC=C1CP(=O)(O)O)O

Specifications

Undesignated
200mg
C7H9O4P
3261
C1=CC(=CC=C1CP(=O)(O)O)O
188.119
188.12
≥98.0% (T)
222°C
90001-07-3
MFCD16877729
ZHBRSHSRMYZHLS-UHFFFAOYSA-N
(4-hydroxyphenyl)methylphosphonic acid
12029255
Crystal-Powder at 20°C
(4-Hydroxybenzyl)phosphonic Acid
Safety and Handling

Safety and Handling

TSCA : No