Learn More
4-Hydroxybenzoic Acid Hydrazide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 121005000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Hydroxybenzoic acid hydrazide | |
97.5 | |
Authentic | |
Plastic bottle | |
HOC6H4CONHNH2 | |
500g | |
4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide | |
ZMZGIVVRBMFZSG-UHFFFAOYSA-N | |
4-hydroxybenzohydrazide | |
1742 | |
98% |
5351-23-5 | |
100.0 | |
98% | |
C7H8N2O2 | |
MFCD00007605 | |
10, 174 | |
Solubility in water: 26.5g/L (25°C). Other solubilities: soluble in methanol,ethanol | |
C1=CC(=CC=C1C(=O)NN)O | |
152.15 | |
152.15 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye pr
GHS Signal Word: Warning
EINECSNumber : 226-326-5
RUO â Research Use Only