Learn More
4-Hydroxybenzenesulfonic acid, 65%, Thermo Scientific Chemicals
$92.82 - $323.84
Chemical Identifiers
CAS | 98-67-9 |
---|---|
Molecular Formula | C6H6O4S |
Molecular Weight (g/mol) | 174.17 |
MDL Number | MFCD00007506 |
InChI Key | FEPBITJSIHRMRT-UHFFFAOYSA-N |
Synonym | p-phenolsulfonic acid, phenolsulfonic acid, 4-phenolsulfonic acid, p-sulfophenol, sulfocarbolic acid, benzenesulfonic acid, 4-hydroxy, p-hydroxybenzenesulfonic acid, phenol-4-sulfonic acid, benzenesulfonic acid, p-hydroxy, 4-hydroxyphenylsulfonic acid |
PubChem CID | 4765 |
ChEBI | CHEBI:32354 |
IUPAC Name | 4-hydroxybenzenesulfonic acid |
SMILES | OC1=CC=C(C=C1)S(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH59917AK
|
Thermo Scientific Chemicals
H59917AK |
250 mL |
Each for $92.82
|
|
|||||
AAH59917K2
|
Thermo Scientific Chemicals
H59917K2 |
1 L |
Each for $323.84
|
|
|||||
Description
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
98-67-9 | |
174.17 | |
FEPBITJSIHRMRT-UHFFFAOYSA-N | |
4765 | |
4-hydroxybenzenesulfonic acid |
C6H6O4S | |
MFCD00007506 | |
p-phenolsulfonic acid, phenolsulfonic acid, 4-phenolsulfonic acid, p-sulfophenol, sulfocarbolic acid, benzenesulfonic acid, 4-hydroxy, p-hydroxybenzenesulfonic acid, phenol-4-sulfonic acid, benzenesulfonic acid, p-hydroxy, 4-hydroxyphenylsulfonic acid | |
CHEBI:32354 | |
OC1=CC=C(C=C1)S(O)(=O)=O |
Specifications
98-67-9 | |
C6H6O4S | |
MFCD00007506 | |
UN1803 | |
p-phenolsulfonic acid, phenolsulfonic acid, 4-phenolsulfonic acid, p-sulfophenol, sulfocarbolic acid, benzenesulfonic acid, 4-hydroxy, p-hydroxybenzenesulfonic acid, phenol-4-sulfonic acid, benzenesulfonic acid, p-hydroxy, 4-hydroxyphenylsulfonic acid | |
FEPBITJSIHRMRT-UHFFFAOYSA-N | |
4-hydroxybenzenesulfonic acid | |
4765 | |
174.17 | |
Liquid |
1.337 g/mL | |
1.489 | |
250 mL | |
1869034 | |
Soluble in water. | |
OC1=CC=C(C=C1)S(O)(=O)=O | |
174.17 | |
CHEBI:32354 | |
65% | |
4-Hydroxybenzenesulfonic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: PHENOLSULFONIC ACID, LIQUID
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only