Please login to your online account to display your discounted pricing

4-Hydroxybenzamide, +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Hydroxybenzamide
CAS 619-57-8
CAS Min % 97.0
CAS Max % 100.0
Assay 97.0% min.
Assay Percent Range 97+%
Molecular Formula C7H7NO2
Linear Formula OHC6H4C(=O)NH2
Formula Weight 137.14g/mol
MDL Number MFCD00007997
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC394110250

  SDS
thermo fisher scientific
394110250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White to off-white
Melting Point 159° to 163°C
Quantity 25g
Chemical Name or Material 4-Hydroxybenzamide
CAS 619-57-8
CAS Min % 97.0
CAS Max % 100.0
Physical Form White to off-white crystalline powder
Assay 97.0% min.
Assay Percent Range 97+%
Molecular Formula C7H7NO2
Linear Formula OHC6H4C(=O)NH2
Formula Weight 137.14g/mol
MDL Number MFCD00007997
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.