missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybenzaldehyde 98.0+%, TCI America™
Supplier: TCI America H0198100G
Specifications
| 4-Hydroxybenzaldehyde | |
| 117°C | |
| MFCD00006939 | |
| p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde | |
| C1=CC(=CC=C1C=O)O | |
| 122.123 | |
| CHEBI:17597 | |
| ≥98.0% (GC) |
| 123-08-0 | |
| C7H6O2 | |
| 100 g | |
| RGHHSNMVTDWUBI-UHFFFAOYSA-N | |
| 4-hydroxybenzaldehyde | |
| 126 | |
| 122.12 | |
| Crystalline Powder |
Chemical Identifiers
| 123-08-0 | |
| 122.123 | |
| RGHHSNMVTDWUBI-UHFFFAOYSA-N | |
| 126 | |
| 4-hydroxybenzaldehyde |
| C7H6O2 | |
| MFCD00006939 | |
| p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde | |
| CHEBI:17597 | |
| C1=CC(=CC=C1C=O)O |
Safety and Handling
EINECSNumber : (3)-1183&(3)-2660
RTECSNumber : CU6475000
TSCA : Yes