missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxybenzaldehyde, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 162770500

 View more versions of this product

Catalog No. AC162770500



Specifications

Specifications

4-Hydroxybenzaldehyde
Plastic bottle
114.0°C to 118.0°C
123-08-0
C7H6O2
08,64
4-formylphenol
RGHHSNMVTDWUBI-UHFFFAOYSA-N
4-hydroxybenzaldehyde
126
122.12
99%
Authentic
Yellow to Brown
50g
98.5% min. (GC)
HOC6H4CHO
15,4849
Solubility in water: 13g/L (30°C). Other solubilities: 36.8g/L benzene (65°C)
C1=CC(=CC=C1C=O)O
122.12
CHEBI:17597
Crystalline Powder

Chemical Identifiers

123-08-0
122.12
4-formylphenol
CHEBI:17597
C1=CC(=CC=C1C=O)O
C7H6O2
RGHHSNMVTDWUBI-UHFFFAOYSA-N
126
4-hydroxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

EINECSNumber : 204-599-1

SDS Documents