missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Hydroxyacetophenone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150405000

 View more versions of this product

Catalog No. AC150405000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Hydroxyacetophenone
99-93-4
100.0
Beige to White
166°C
98%
C8H8O2
MFCD00002359
08, 87
Solubility in water: 10g/L (22°C)
CC(=O)C1=CC=C(O)C=C1
136.15
CHEBI:28032
98%
98%
97.5
107°C to 111°C
147°C to 148°C (3.0 mmHg)
Authentic
Plastic bottle
HOC6H4COCH3
500 g
4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone
TXFPEBPIARQUIG-UHFFFAOYSA-N
1-(4-hydroxyphenyl)ethanone
7469
136.15
Crystalline Powder

Chemical Identifiers

99-93-4
136.15
TXFPEBPIARQUIG-UHFFFAOYSA-N
7469
1-(4-hydroxyphenyl)ethanone
C8H8O2
MFCD00002359
4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone
CHEBI:28032
CC(=O)C1=CC=C(O)C=C1
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-802-8

RTECSNumber : AM8750000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only