missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-7-methoxycoumarin 98.0+%, TCI America™

$181.05 - $536.56

Chemical Identifiers

CAS 17575-15-4
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00673700
InChI Key MJBHLQMPKBMZSF-UHFFFAOYSA-N
Synonym 2-hydroxy-7-methoxy-4h-chromen-4-one
PubChem CID 54691408
IUPAC Name 4-hydroxy-7-methoxychromen-2-one
SMILES COC1=CC2=C(C=C1)C(=CC(=O)O2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14201G View Documents Tci America
H14201G
1g Each for $181.05
 
H14205G View Documents Tci America
H14205G
5g Each for $536.56
 
Specifications

Chemical Identifiers

17575-15-4
192.17
MJBHLQMPKBMZSF-UHFFFAOYSA-N
54691408
COC1=CC2=C(C=C1)C(=CC(=O)O2)O
C10H8O4
MFCD00673700
2-hydroxy-7-methoxy-4h-chromen-4-one
4-hydroxy-7-methoxychromen-2-one

Specifications

Undesignated
1g
C10H8O4
2-hydroxy-7-methoxy-4h-chromen-4-one
COC1=CC2=C(C=C1)C(=CC(=O)O2)O
192.17
192.17
≥98.0% (GC,T)
260°C
17575-15-4
MFCD00673700
MJBHLQMPKBMZSF-UHFFFAOYSA-N
4-hydroxy-7-methoxychromen-2-one
54691408
Crystal-Powder at 20°C
4-Hydroxy-7-methoxycoumarin
Safety and Handling

Safety and Handling

TSCA : No