Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-6-(trifluoromethyl)quinoline, 98%

$329.99 - $329.99

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-6-(trifluoromethyl)quinoline
CAS 49713-51-1
Molecular Formula C10H6F3NO
Formula Weight 213.16
MDL Number MFCD00209673
Synonym 6-Trifluoromethyl-4-quinolinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1855506

  SDS
alfa aesar™
A1855506
5g Each for $329.99
Description & Specifications

Specifications

Formula Weight 213.16
Chemical Name or Material 4-Hydroxy-6-(trifluoromethyl)quinoline
MDL Number MFCD00209673
Assay 98%
Molecular Formula C10H6F3NO
CAS 49713-51-1
Synonym 6-Trifluoromethyl-4-quinolinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 265° to 268°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.