missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-6-(trifluoromethyl)pyrimidine, 97%

$37.46 - $176.45

Chemical Identifiers

CAS 1546-78-7
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.087
MDL Number MFCD00192530
InChI Key LVOYSBZJJWPUBD-UHFFFAOYSA-N
Synonym 4 1h-pyrimidinone, 6-trifluoromethyl, 4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-4-hydroxypyrimidine, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-pyrimidin-4-ol
PubChem CID 1268071
IUPAC Name 6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES C1=C(NC=NC1=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6438103 View Documents Alfa Aesar™
H6438103
1g Each for $37.46
 
AAH6438106 View Documents Alfa Aesar™
H6438106
5g Each for $176.45
 
Description

Description

Applications
6-(Trifluoromethyl)-4-pyrimidinol is used as building block in chemical synthesis. It may also be used in the synthesis of 10-[3-(6-trifluoromethyl-pyrimidin-4-yloxy)-propoxy]-(20S)-camptothecin.

Solubility
Solubility in methanol (almost transparency). Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat.
Specifications

Chemical Identifiers

1546-78-7
164.087
LVOYSBZJJWPUBD-UHFFFAOYSA-N
1268071
C1=C(NC=NC1=O)C(F)(F)F
C5H3F3N2O
MFCD00192530
4 1h-pyrimidinone, 6-trifluoromethyl, 4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-4-hydroxypyrimidine, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-pyrimidin-4-ol
6-(trifluoromethyl)-1H-pyrimidin-4-one

Specifications

White
1g
C5H3F3N2O
145548
LVOYSBZJJWPUBD-UHFFFAOYSA-N
6-(trifluoromethyl)-1H-pyrimidin-4-one
1268071
Crystalline
4-Hydroxy-6-(trifluoromethyl)pyrimidine
169°C to 171°C
1546-78-7
MFCD00192530
4 1h-pyrimidinone, 6-trifluoromethyl, 4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-4-hydroxypyrimidine, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-pyrimidin-4-ol
C1=C(NC=NC1=O)C(F)(F)F
164.087
164.09
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures