missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hydroxy-6-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6438106
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications6-(Trifluoromethyl)-4-pyrimidinol is used as building block in chemical synthesis. It may also be used in the synthesis of 10-[3-(6-trifluoromethyl-pyrimidin-4-yloxy)-propoxy]-(20S)-camptothecin.
Solubility
Solubility in methanol (almost transparency). Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat.
Specifications
4-Hydroxy-6-(trifluoromethyl)pyrimidine | |
169°C to 171°C | |
C5H3F3N2O | |
5 g | |
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl | |
C1=C(NC=NC1=O)C(F)(F)F | |
164.087 | |
164.09 | |
Crystalline |
1546-78-7 | |
White | |
MFCD00192530 | |
145548 | |
LVOYSBZJJWPUBD-UHFFFAOYSA-N | |
6-(trifluoromethyl)-1H-pyrimidin-4-one | |
1268071 | |
97% |
Chemical Identifiers
1546-78-7 | |
164.087 | |
LVOYSBZJJWPUBD-UHFFFAOYSA-N | |
1268071 | |
C1=C(NC=NC1=O)C(F)(F)F |
C5H3F3N2O | |
MFCD00192530 | |
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl | |
6-(trifluoromethyl)-1H-pyrimidin-4-one |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures