Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol, 97+%

$56.95 - $628.32

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-6- (trifluoromethyl)pyrimidine-2-thiol
CAS 368-54-7
Molecular Formula C5H3F3N2OS
Formula Weight 196.1
MDL Number MFCD00112978
Synonym 4-Hydroxy-2-mercapto-6-(trifluoromethyl)pyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4387302

  SDS
alfa aesar™
4387302
0.5g Each for $56.95

AA4387309

  SDS
alfa aesar™
4387309
10g Each for $628.32
Description & Specifications

Specifications

Formula Weight 196.1
Chemical Name or Material 4-Hydroxy-6- (trifluoromethyl)pyrimidine-2-thiol
Physical Form Powder
Assay ≥97%
Molecular Formula C5H3F3N2OS
MDL Number MFCD00112978
CAS 368-54-7
Synonym 4-Hydroxy-2-mercapto-6-(trifluoromethyl)pyrimidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 251°C
Quantity 0.5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.