Please login to your online account to display your discounted pricing

4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid
CAS 175203-86-8
Assay Tech.
Assay Percent Range 97%
Molecular Formula C11H6F3NO4
Formula Weight 273.17
MDL Number MFCD00219848
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM09299DA

  SDS
fisher scientific company
KM09299DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM09299EA

  SDS
fisher scientific company
KM09299EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 273.17
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Chemical Name or Material 4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid
Molecular Formula C11H6F3NO4
CAS 175203-86-8
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00219848
Packaging Amber glass bottle
Assay Percent Range 97%