Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-6-methylpyrimidine, 97%

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-6-methylpyrimidine
CAS 3524-87-6
Formula Weight 110.11
MDL Number MFCD00053575
Synonym 6-Methyl-4-pyrimidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6140403

 
alfa aesar™
H6140403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 110.11
Melting Point 145° to 150°C
Chemical Name or Material 4-Hydroxy-6-methylpyrimidine
TSCA TSCA
Physical Form Solid
Assay 97%
MDL Number MFCD00053575
CAS 3524-87-6
Synonym 6-Methyl-4-pyrimidinol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UW7528000
Color White to brown
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.