missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-6-methylcoumarin 98.0+%, TCI America™

$74.06 - $245.23

Chemical Identifiers

CAS 13252-83-0
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00085738
InChI Key WIRGBZBGYNIZIB-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methyl-4h-chromen-4-one, 2-hydroxy-6-methylchromen-4-one, 2h-1-benzopyran-2-one, 4-hydroxy-6-methyl, 2h-1-benzopyran-2-one,4-hydroxy-6-methyl, 4-hydroxy-6-methyl-2h-chromen-2-one, 4-hydroxy-6-methylcoumarin, 6-methyl-4-hydroxy coumarin, 6-methyl-4-hydroxycoumarin, hydroxy-6-methylcoumarin,4
PubChem CID 54686349
IUPAC Name 4-hydroxy-6-methylchromen-2-one
SMILES CC1=CC2=C(C=C1)OC(=O)C=C2O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14211G View Documents Tci America
H14211G
1g Each for $74.06
 
H14215G View Documents Tci America
H14215G
5g Each for $245.23
 
Specifications

Chemical Identifiers

13252-83-0
176.171
WIRGBZBGYNIZIB-UHFFFAOYSA-N
54686349
CC1=CC2=C(C=C1)OC(=O)C=C2O
C10H8O3
MFCD00085738
2-hydroxy-6-methyl-4h-chromen-4-one, 2-hydroxy-6-methylchromen-4-one, 2h-1-benzopyran-2-one, 4-hydroxy-6-methyl, 2h-1-benzopyran-2-one,4-hydroxy-6-methyl, 4-hydroxy-6-methyl-2h-chromen-2-one, 4-hydroxy-6-methylcoumarin, 6-methyl-4-hydroxy coumarin, 6-methyl-4-hydroxycoumarin, hydroxy-6-methylcoumarin,4
4-hydroxy-6-methylchromen-2-one

Specifications

White
1g
C10H8O3
2-hydroxy-6-methyl-4h-chromen-4-one, 2-hydroxy-6-methylchromen-4-one, 2h-1-benzopyran-2-one, 4-hydroxy-6-methyl, 2h-1-benzopyran-2-one,4-hydroxy-6-methyl, 4-hydroxy-6-methyl-2h-chromen-2-one, 4-hydroxy-6-methylcoumarin, 6-methyl-4-hydroxy coumarin, 6-methyl-4-hydroxycoumarin, hydroxy-6-methylcoumarin,4
CC1=CC2=C(C=C1)OC(=O)C=C2O
176.171
176.17
≥98.0% (GC)
261°C
13252-83-0
MFCD00085738
WIRGBZBGYNIZIB-UHFFFAOYSA-N
4-hydroxy-6-methylchromen-2-one
54686349
Crystal-Powder at 20°C
4-Hydroxy-6-methylcoumarin
Safety and Handling

Safety and Handling

TSCA : No