missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-6-methyl-3-nitro-2-pyridone, 98+%, ACROS Organics™

Chemical Identifiers

CAS 4966-90-9
Molecular Formula C6H6N2O4
Molecular Weight (g/mol) 170.13
MDL Number MFCD00167612
InChI Key QIKWTNPFTOEELW-UHFFFAOYSA-N
Synonym 2,4-dihydroxy-6-methyl-3-nitropyridine
PubChem CID 54685619
IUPAC Name 4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one
SMILES CC1=CC(=C(C(=O)N1)[N+](=O)[O-])O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367820050 SDS View Documents Thermo Fisher Scientific
AC367820050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC367820250 SDS View Documents Thermo Fisher Scientific
AC367820250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

4966-90-9
170.13
QIKWTNPFTOEELW-UHFFFAOYSA-N
54685619
CC1=CC(=C(C(=O)N1)[N+](=O)[O-])O
C6H6N2O4
MFCD00167612
2,4-dihydroxy-6-methyl-3-nitropyridine
4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one

Specifications

Authentic
Yellow
5g
98.0
98% min. (HPLC)
MFCD00167612
QIKWTNPFTOEELW-UHFFFAOYSA-N
4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one
54685619
Powder
4-Hydroxy-6-methyl-3-nitro-2-pyridone
Glass bottle
293.0°C to 296.0°C
4966-90-9
100.0
C6H6N2O4
2,4-dihydroxy-6-methyl-3-nitropyridine
CC1=CC(=C(C(=O)N1)[N+](=O)[O-])O
170.13
170.13
98+%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning