missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-6-methyl-3-nitro-2-pyridone, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC367820250

 View more versions of this product

Catalog No. AC367820250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Hydroxy-6-methyl-3-nitro-2-pyridone
Glass bottle
293.0°C to 296.0°C
4966-90-9
100.0
C6H6N2O4
2,4-dihydroxy-6-methyl-3-nitropyridine, 2-hydroxy-6-methyl-3-nitro-1h-pyridin-4-one, 4-hydroxy-6-methyl-3-nitro-1,2-dihydropyridin-2-one, 4-hydroxy-6-methyl-3-nitro-2 1h-pyridinone, 4-hydroxy-6-methyl-3-nitro-2-pyridone, 4-hydroxy-6-methyl-3-nitrohydropyridin-2-one, 4-hydroxy-6-methyl-3-nitropyridin-2 1h-one, 6-methyl-3-nitropyridine-2,4-diol, acmc-209khe, pubchem3892
CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O
170.12
170.13
98+%
Authentic
Yellow
25g
98.0
98% min. (HPLC)
MFCD00167612
QIKWTNPFTOEELW-UHFFFAOYSA-N
4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one
54685619
Powder

Chemical Identifiers

4966-90-9
170.12
QIKWTNPFTOEELW-UHFFFAOYSA-N
54685619
CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O
C6H6N2O4
MFCD00167612
2,4-dihydroxy-6-methyl-3-nitropyridine, 2-hydroxy-6-methyl-3-nitro-1h-pyridin-4-one, 4-hydroxy-6-methyl-3-nitro-1,2-dihydropyridin-2-one, 4-hydroxy-6-methyl-3-nitro-2 1h-pyridinone, 4-hydroxy-6-methyl-3-nitro-2-pyridone, 4-hydroxy-6-methyl-3-nitrohydropyridin-2-one, 4-hydroxy-6-methyl-3-nitropyridin-2 1h-one, 6-methyl-3-nitropyridine-2,4-diol, acmc-209khe, pubchem3892
4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents