Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-6-methyl-2-(trifluoromethyl)quinoline, 97%

$52.43 - $207.93

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-6-methyl-2-(trifluoromethyl)quinoline
CAS 1701-20-8
Molecular Formula C11H8F3NO
Formula Weight 227.19
MDL Number MFCD00153192
Synonym 6-Methyl-2-(trifluoromethyl)-4-quinolinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2067503

  SDS
alfa aesar™
B2067503
1g Each for $52.43

AAB2067506

  SDS
alfa aesar™
B2067506
5g Each for $207.93
Description & Specifications

Specifications

Formula Weight 227.19
Chemical Name or Material 4-Hydroxy-6-methyl-2-(trifluoromethyl)quinoline
Assay 97%
Molecular Formula C11H8F3NO
MDL Number MFCD00153192
CAS 1701-20-8
Synonym 6-Methyl-2-(trifluoromethyl)-4-quinolinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 252° to 253°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.