Please login to your online account to display your discounted pricing

4-Hydroxy-6-methoxymethyl-2-(methylthio)pyrimidine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RJF00491DA

 View more versions of this product

Catalog No. RJF00491DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 186.23
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-Hydroxy-6-methoxymethyl-2-(methylthio)pyrimidine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00052618
Packaging Amber Glass Bottle
Molecular Formula C7H10N2O2S
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Assay Percent Range 97%
CAS 68087-13-8

Specifications

Chemical Name or Material 4-Hydroxy-6-methoxymethyl-2-(methylthio)pyrimidine
CAS 68087-13-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2O2S
Formula Weight 186.23
MDL Number MFCD00052618
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents