Please login to your online account to display your discounted pricing

4-hydroxy-6-mercaptopyrazolo[3,4-d]pyrimidine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-6-mercaptopyrazolo[3, 4-d]pyrimidine
CAS 24521-76-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H4N4OS
Formula Weight 168.18
MDL Number MFCD00022782
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376630010

  SDS
thermo fisher scientific
AC376630010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376630050

  SDS
thermo fisher scientific
AC376630050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 168.18
Quantity 1g
Color Light yellow
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00022782
CAS 24521-76-4
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 4-Hydroxy-6-mercaptopyrazolo[3, 4-d]pyrimidine
Molecular Formula C5H4N4OS
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.