Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine, 98%

$148.18 - $148.18

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine
CAS 24521-76-4
Molecular Formula C5H4N4OS
Formula Weight 168.18g/mol
MDL Number MFCD00022782
Beilstein 140413
Synonym 1,5,6,7-Tetrahydro-6-thioxo-4H-pyrazolo[3,4-d]pyrimidin-4-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0683603

 
alfa aesar™
L06836-03
1g Each for $148.18
Description & Specifications

Specifications

Melting Point >310°C
Quantity 1g
Chemical Name or Material 4-Hydroxy-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine
CAS 24521-76-4
Assay 98%
Molecular Formula C5H4N4OS
Formula Weight 168.18g/mol
MDL Number MFCD00022782
Beilstein 140413
Synonym 1,5,6,7-Tetrahydro-6-thioxo-4H-pyrazolo[3,4-d]pyrimidin-4-one