missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-5-iodopyrimidine, 95%

Manufacturer:  Alfa Aesar™ H61190MD

 View more versions of this product

Catalog No. AAH61190MD



Description

Description

Applications
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

Solubility
Soluble in water (67 g/L) (25°C).

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

4-Hydroxy-5-iodopyrimidine
250mg
C11H12N2O3
2s-2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid
C1=CC2=C(C=C1O)C(=CN2)CC(C(=O)O)N
220.23
CHEBI:17780
Solid
Light Sensitive
4350-09-8
MFCD02666079
LDCYZAJDBXYCGN-VIFPVBQESA-N
(2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
439280
221.98
95%

Chemical Identifiers

4350-09-8
220.23
LDCYZAJDBXYCGN-VIFPVBQESA-N
439280
(2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
C11H12N2O3
MFCD02666079
2s-2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid
CHEBI:17780
C1=CC2=C(C=C1O)C(=CN2)CC(C(=O)O)N
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents