Learn More
4-Hydroxy-4-methyl-2-pentanone, 98+%, Thermo Scientific Chemicals
$39.85 - $262.58
Chemical Identifiers
CAS | 123-42-2 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00004471 |
InChI Key | SWXVUIWOUIDPGS-UHFFFAOYSA-N |
Synonym | diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one |
PubChem CID | 31256 |
ChEBI | CHEBI:55381 |
IUPAC Name | 4-hydroxy-4-methylpentan-2-one |
SMILES | CC(=O)CC(C)(C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA16248AE
|
Thermo Scientific Chemicals
A16248AE |
100 mL |
N/A
|
|
|||||
AAA16248AP
|
Thermo Scientific Chemicals
A16248AP |
500 mL |
N/A
|
|
|||||
AAA162480F
|
Thermo Scientific Chemicals
A162480F |
2500 mL |
N/A
|
|
|||||
Description
4-Hydroxy-4-methyl-2-pentanone, is used as a solvent. It is also used as an intermediate of chemical products and is used as solvent for some industrial paint to adjust thesolubility of paint resins and regulate the evaporation rate. It is mainly used in paints and coatings (coil coatings, wood varnishes, architectural coatings…). It is a good solvent for organic peroxides.It can also be used for the treatment of textiles and leather, in chemical synthesis or as a cleaning solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxy-4-methyl-2-pentanone, is used as a solvent. It is also used as an intermediate of chemical products and is used as solvent for some industrial paint to adjust thesolubility of paint resins and regulate the evaporation rate. It is mainly used in paints and coatings (coil coatings, wood varnishes, architectural coatingsâ¦). It is a good solvent for organic peroxides.It can also be used for the treatment of textiles and leather, in chemical synthesis or as a cleaning solvent.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases.
Chemical Identifiers
123-42-2 | |
116.16 | |
SWXVUIWOUIDPGS-UHFFFAOYSA-N | |
31256 | |
4-hydroxy-4-methylpentan-2-one |
C6H12O2 | |
MFCD00004471 | |
diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one | |
CHEBI:55381 | |
CC(=O)CC(C)(C)O |
Specifications
123-42-2 | |
0.935 | |
58°C (136°F) | |
C6H12O2 | |
MFCD00004471 | |
UN1148 | |
14,2964 | |
It is soluble in water. | |
CC(=O)CC(C)(C)O | |
116.16 | |
CHEBI:55381 | |
≥98% |
-47°C | |
168°C | |
Pleasant | |
1.423 | |
100 mL | |
1740440 | |
diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one | |
SWXVUIWOUIDPGS-UHFFFAOYSA-N | |
4-hydroxy-4-methylpentan-2-one | |
31256 | |
116.16 | |
4-Hydroxy-4-methyl-2-pentanone |
Safety and Handling
GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
H226-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIACETONE ALCOHOL
EINECSNumber : 204-626-7
RTECSNumber : SA9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only