Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-4'-iodobiphenyl, 98+%

$845.30 - $845.30

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-4'-iodobiphenyl
CAS 29558-78-9
Molecular Formula C12H9IO
Formula Weight 296.11
MDL Number MFCD02258843
Synonym 4-(4-Iodophenyl)phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2211522

  SDS
alfa aesar™
B2211522
100g Each for $845.30
Description & Specifications

Specifications

Formula Weight 296.11
MDL Number MFCD02258843
Molecular Formula C12H9IO
Synonym 4-(4-Iodophenyl)phenol
Assay ≥98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Hydroxy-4'-iodobiphenyl
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 29558-78-9
Melting Point 117° to 119°C
Health Hazard 1 Warning
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.