Please login to your online account to display your discounted pricing

4'-Hydroxy-4-biphenylcarboxylic acid, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338370050

 View more versions of this product

Catalog No. AC338370050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (HPLC)
Physical Form Liquid or Solid
Formula Weight 214.22
Quantity 5g
Color Clear colorless to yellow
TSCA TSCA
Melting Point 295° to 305°C
CAS Max % 100.0
Chemical Name or Material 4'-Hydroxy-4-biphenylcarboxylic acid
Packaging Glass bottle
Name Note 98%
Assay Percent Range 99%
Molecular Formula C13H10O3
Linear Formula HOC6H4C6H4COOH
MDL Number MFCD00059078
CAS 58574-03-1
Synonym 4-(4-Hydroxyphenyl)benzoic acid
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4'-Hydroxy-4-biphenylcarboxylic acid
Name Note 98%
CAS 58574-03-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥97.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C13H10O3
Linear Formula HOC6H4C6H4COOH
Formula Weight 214.22
MDL Number MFCD00059078
Synonym 4-(4-Hydroxyphenyl)benzoic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents