missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Hydroxy-4-biphenylcarboxylic acid, 99%, ACROS Organics™

Chemical Identifiers

CAS 58574-03-1
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C13H10O3
Linear Formula HOC6H4C6H4COOH
Formula Weight 214.22
MDL Number MFCD00059078
Synonym 4-(4-Hydroxyphenyl)benzoic acid
Chemical Name or Material 4'-Hydroxy-4-biphenylcarboxylic acid, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC338370050 SDS Thermo Fisher Scientific
338370050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

58574-03-1
100.0
C13H10O3
214.22
4-(4-Hydroxyphenyl)benzoic acid
≥97.5% (HPLC)
98.5
99%
HOC6H4C6H4COOH
MFCD00059078
4'-Hydroxy-4-biphenylcarboxylic acid, 98%

Specifications

Clear colorless to yellow
5g
98.5
Authentic
Liquid or Solid
C13H10O3
214.22
4-(4-Hydroxyphenyl)benzoic acid
≥97.5% (HPLC)
295° to 305°C
58574-03-1
100.0
Glass bottle
99%
HOC6H4C6H4COOH
MFCD00059078
4'-Hydroxy-4-biphenylcarboxylic acid, 98%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA