Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-(trifluoromethyl)benzaldehyde, 98%

$169.32 - $429.46

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-(trifluoromethyl)benzaldehyde
CAS 220227-98-5
Molecular Formula C8H5F3O2
Formula Weight 190.12
MDL Number MFCD01091006
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2609803

  SDS
alfa aesar™
H2609803
1g Each for $429.46

AAH26098MD

  SDS
alfa aesar™
H26098MD
250mg Each for $169.32
Description & Specifications

Specifications

Formula Weight 190.12
Assay 98%
Molecular Formula C8H5F3O2
MDL Number MFCD01091006
CAS 220227-98-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 159° to 163°C
Quantity 1g
Chemical Name or Material 4-Hydroxy-3-(trifluoromethyl)benzaldehyde
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.