Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-oxo-2,5-diphenyl-2,3-dihydrothiophene 1,1-dioxide, 98%

$71.27 - $209.45

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-oxo-2,5-diphenyl-2,3-dihydrothiophene 1,1-dioxide
CAS 54714-10-2
Molecular Formula C16H12O4S
Formula Weight 300.34
MDL Number MFCD00075268
Beilstein 544870
Synonym HOTDO; 2,5-Diphenyl-4-hydroxy-3-oxo-2,3-dihydrothiophene 1,1-dioxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2415306

  SDS
alfa aesar™
B2415306
5g Each for $71.27

AAB2415314

  SDS
alfa aesar™
B2415314
25g Each for $209.45
Description & Specifications

Specifications

Formula Weight 300.34
Chemical Name or Material 4-Hydroxy-3-oxo-2,5-diphenyl-2,3-dihydrothiophene 1,1-dioxide
Assay 98%
Molecular Formula C16H12O4S
MDL Number MFCD00075268
CAS 54714-10-2
Beilstein 544870
Synonym HOTDO; 2,5-Diphenyl-4-hydroxy-3-oxo-2,3-dihydrothiophene 1,1-dioxide
Melting Point 238° to 241°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.