missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-3-nitropyridine, TCI America™

$84.03 - $270.42

Chemical Identifiers

CAS 5435-54-1
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
InChI Key YUWOLBZMQDGRFV-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitropyridine, 3-nitropyridin-4-ol, 3-nitro-4-pyridinol, 3-nitro-4-hydroxypyridine, 3-nitropyridin-4 1h-one, 3-nitro-4-pyridone, 4-pyridinol, 3-nitro, 4 1h-pyridinone, 3-nitro, 3-nitro-1,4-dihydropyridin-4-one, 4-hydroxy-3-nitropytidine
PubChem CID 228506
IUPAC Name 3-nitro-1H-pyridin-4-one
SMILES C1=CNC=C(C1=O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H117125G View Documents TCI America
H117125G
25g Each for $270.42
 
H11715G View Documents TCI America
H11715G
5g Each for $84.03
 
Specifications

Chemical Identifiers

5435-54-1
140.098
4-hydroxy-3-nitropyridine, 3-nitropyridin-4-ol, 3-nitro-4-pyridinol, 3-nitro-4-hydroxypyridine, 3-nitropyridin-4 1h-one, 3-nitro-4-pyridone, 4-pyridinol, 3-nitro, 4 1h-pyridinone, 3-nitro, 3-nitro-1,4-dihydropyridin-4-one, 4-hydroxy-3-nitropytidine
3-nitro-1H-pyridin-4-one
C5H4N2O3
YUWOLBZMQDGRFV-UHFFFAOYSA-N
228506
C1=CNC=C(C1=O)[N+](=O)[O-]

Specifications

Undesignated
25g
C5H4N2O3
YUWOLBZMQDGRFV-UHFFFAOYSA-N
3-nitro-1H-pyridin-4-one
228506
Crystal-Powder at 20°C
4-Hydroxy-3-nitropyridine
284°C
5435-54-1
4-hydroxy-3-nitropyridine, 3-nitropyridin-4-ol, 3-nitro-4-pyridinol, 3-nitro-4-hydroxypyridine, 3-nitropyridin-4 1h-one, 3-nitro-4-pyridone, 4-pyridinol, 3-nitro, 4 1h-pyridinone, 3-nitro, 3-nitro-1,4-dihydropyridin-4-one, 4-hydroxy-3-nitropytidine
C1=CNC=C(C1=O)[N+](=O)[O-]
140.098
140.10
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No